Learn More
Azure Biosystems chemiSOLO Imaging System
Azure Biosystems chemiSOLO Imaging System is a chemiluminescence and visible gel imaging system for quantitative imaging of Western blots and visible gels.
Supplier:  Azure Biosystems CS1000
Catalog No. CS1000AZ
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Compact chemiluminescent imager for quantitative chemiluminescent and visible protein gel imaging
  • Rapidly images chemiluminescent Western blots with femtogram sensitivity
  • Saves benchspace with compact design, compared to entire darkrooms and bulky imagers
  • Allows for Western blot quantitation over a wide dynamic range using sensitive, linear detection and advanced image processing like Extended Dynamic Range exposure
  • Captures colorimetric blots and visible-stained protein gels in addition to chemiluminescent detection, allowing for gel documentation and chemi marker functionality
Specifications
Specifications
None
Chemiluminescence Imaging
Imaging System
Product Suggestions
Product Suggestions