Learn More
Azure Biosystems chemiSOLO and Densitometry Kit Package
Description
- Compact chemiluminescent imager for quantitative chemiluminescent and visible protein gel imaging
- Rapidly images chemiluminescent Western blots with femtogram sensitivity
- Saves benchspace with compact design, compared to entire darkrooms and bulky imagers
- Allows for Western blot quantitation over a wide dynamic range using sensitive, linear detection and advanced image processing like Extended Dynamic Range exposure
- Captures colorimetric blots and visible-stained protein gels in addition to chemiluminescent detection, allowing for gel documentation and chemi marker functionality
- Densitometry Kit provides materials for densitometric verification and analysis of visible-stained gels
- Kit includes: 21-Step Densitometry Calibration Target and chemiSOLO Densitometry Protocol
Specifications
Specifications
| For Use With (Application) | Chemiluminescence Imaging |
| Product Type | Imaging System |
| Display Type | None |