Learn More
Azure Biosystems chemiSOLO and Chemi Western Blot Starter Kit Package
Azure Biosystems chemiSOLO and Chemi Western Blot Starter Package includes both the chemiSOLO imaging system and the Chemi Western Blot Starter Kit, a collection of Western blotting materials for labs new to chemiluminescent Western blotting.
Supplier:  Azure Biosystems CS1002
Catalog No. CS1002AZ
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Compact chemiluminescent imager for quantitative chemiluminescent and visible protein gel imaging
  • Rapidly images chemiluminescent Western blots with femtogram sensitivity
  • Saves benchspace with compact design, compared to entire darkrooms and bulky imagers
  • Allows for Western blot quantitation over a wide dynamic range using sensitive, linear detection and advanced image processing like Extended Dynamic Range exposure
  • Captures colorimetric blots and visible-stained protein gels in addition to chemiluminescent detection, allowing for gel documentation and chemi marker functionality
  • Chemi Western Blot Starter Kit provides a collection of Western blotting materials to jumpstart labs new to chemiluminescent Western blotting
  • Kit includes: Azure Aqua Quad Mini-Cell, Aqua Transfer Cell, Aqua Power Supply, Radiance Q, LF PVDF Membranes, Azure Chemi Blot Blocking Buffer, Azure Blot Washing Buffer, Azure Transfer Buffer, Incubation Tray (11x9 cm), Quenching Sheets (9x12.5 cm), Blotting Paper for Western Blot (7x9 cm), and Blot Development Folder
Specifications
Specifications
None
Chemiluminescence Imaging
Imaging System
Product Suggestions
Product Suggestions