Learn More
MilliporeSigma™ Chemicon™ Mouse Cortical Neural Stem Cell Expansion Kit
Used in Immunocytochemistry, Stem cell culture applications. MilliporeSigma™; Chemicon™ Mouse Cortical Neural Stem Cell Expansion Kit provides a multi-component system for the culture and analysis of neural stem cells and their differentiated progenies.
Supplier: MilliporeSigma™ SCR032
Description
- Includes mouse cortical neural stem cells, isolated from the cortices of embryonic day 15-18 (E15-E18) C57/BL6 mice, neural stem cell expansion medium, and a panel of antibodies for immunocytochemical staining of neural stem/progenitor cells (Nestin and Sox2) and differentiated neural phenotypes (Map2ab for neurons, GFAP for astrocytes, and O1 for oligodendrocytes)
Specifications
|
|
| Stem Cell Expansion Media | |
| 1 Kit |
| Neural Stem Cells | |
| Immunocytochemistry, Stem cell culture | |
| RUO |
For Research Use Only.