missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Chemically Defined Lipid Concentrate
GREENER_CHOICE

Catalog No. 11905031
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11905031 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11905031 Supplier Gibco™ Supplier No. 11905031
Only null left

Chemically Defined Lipid Concentrate

Insect cell culture tested.

Chemically Defined Lipid Concentrate is a concentrated lipid emulsion designed to reduce or replace fetal bovine serum in cell culture media for a wide variety of applications, including growth and maintenance of CHO, hybridoma, and insect cells in culture; monoclonal antibody production by hybridomas; and viral expression in insect cells. The components of this medium supplement are chemically-defined.

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Cell Type Mammalian Cells
Form Liquid
Product Type Lipid Concentrate
pH 4 to 6
Quantity 100 mL
Shipping Condition Room Temperature
Do you have stability/degradation information for Chemically Defined Lipid Concentrate (Cat. No. 11905031)? How has the product been filtered?

The shelf life of the product in the closed bottle is 12 months from the date of manufacture. In order to prevent “air encounters”, we recommend keeping the head space in the bottle as small as possible. You may consider an Argon overlay as well. Argon being heavier than air will settle on top of the chemical. We recommend dividing the contents of the bottle (100 mL) in usable aliquots ideally such that each aliquot is used up in one application.

The product is filtered with a 0.22 µm Durapore filter or Cartridge from EMD Millipore (Cat. No. MSP000812).

For Research Use Only. Not for use in diagnostic procedures.