missing translation for 'onlineSavingsMsg'
Learn More

Chemical Solution, Buffer Solution pH 10.0 at 25°C, Color Coded Blue, Traceable to NIST, LabChem

Catalog No. p-200003991
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125051 500 mL
LC125054 4 L
LC125055 20 L
LC125052 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC125051 Supplier LabChem Supplier No. LC125051
Only null left

This Chemical Solution, Buffer Solution has a pH of 10.0 at 25°C, is Color Coded Blue, and is Traceable to NIST.

  • Chemical Solution, Buffer Solution.
  • pH 10.0 at 25°C.
  • Color Coded Blue.
  • Traceable to NIST.

Specifications

CAS 26628-22-8, 1310-73-2, 497-19-8, 7732-18-5, 144-55-8
Color Blue
Certificate of Analysis Yes
For Use With (Application) pH Measurement
For Use With (Equipment) pH Meter and Electrode
pH 10
Physical Form Aqueous Solution
Quantity 500 mL
Solution Type Buffer
Packaging Poly
Sterility Non-sterile
Traceability to NIST Yes
Grade Laboratory
Show More Show Less