missing translation for 'onlineSavingsMsg'
Learn More

Chelerythrine chloride, 99+%

Catalog No. AAJ62906LB0
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62906LB0 5 mg
AAJ62906MA 10 mg
AAJ62906MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62906LB0 Supplier Thermo Scientific Chemicals Supplier No. J62906LB0

CAS: 3895-92-9 | C21H18ClNO4 | 383.83 g/mol

A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A natural benzophenanthridine alkaloid
  • Inhibits protein kinase C
  • Induces apoptosis in leukemia cells

Chemical Identifiers

CAS 3895-92-9
Molecular Formula C21H18ClNO4
Molecular Weight (g/mol) 383.83
MDL Number MFCD00060717
InChI Key WEEFNMFMNMASJY-UHFFFAOYSA-M
Synonym chelerythrine chloride, chelerythrine hydrochloride, chelerythrinechloride, unii-7ic98tz0pz, 1,2-dimethoxy-12-methyl-1,3 dioxolo 4',5':4,5 benzo 1,2-c phenanthridin-12-ium chloride, chelerythrine, chloride, 7ic98tz0pz, 1,2-dimethoxy-n-methyl-1,3-benzodioxolo 5,6c phenanthridinium, 1,2-dimethoxy-12-methyl 1,3 benzodioxolo 5,6-c phenanthridinium chloride
PubChem CID 72311
SMILES [Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC

Specifications

Color Yellow to Orange
Quantity 5 mg
Merck Index 14,2051
Solubility Information Soluble in DMSO at 2mg/ml
IUPAC Name 1,2-dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridin-12-ium;chloride
Formula Weight 383.82
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Chelerythrine chloride
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 223-444-9
RTECSNumber FL9200000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only