missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chebulinic acid, ≥90.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 18942-26-2 |
| Linear Formula | C41H32O27 |
| Synonym | Eutannin; NSC 69862 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 956.68 |
| Percent Purity | ≥90.0% (HPLC) |
| Grade | Analytical Standard |