missing translation for 'onlineSavingsMsg'
Learn More

Chebulagic acid, ≥85% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013610
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013610 Supplier Merck Emd Millipore Supplier No. 6945510MG
Only null left
Featured Industries: Food and Beverages

Specifications

CAS 23094-71-5
Physical Form Neat
Linear Formula C41H30O27
Type Analytical Standard
Quantity 10 mg
Formula Weight 954.66
Percent Purity ≥85% (HPLC)
Grade Analytical Standard