missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHD4 Recombinant Human Monoclonal Antibody (K49015_15E2), Invitrogen™
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555994
Description
Product is compatible with indirect ELISA (1 μg/mL, capture antibody). Sequence of this protein is as follows: EMGEEEEVER EIIKQEESVD PDYWEKLLRH HYEQQQEDLA RNLGKGKRIR KQVNYNDGSQ EDRDWQDDQS DNQSDYSVAS EEGDEDFDER SEAPRRPSRK GLRNDKDKPL PPLLARVGGN IEVLGFNARQ RKAFLNAIMR YGMPPQDAFT TQWLVRDLRG KSEKEFKAYV SLFMRHLCEP GADGAETFAD GVPREGLSRQ HVLTRIGVMS LIRKKVQEFE HVNGRWSMPE LAEVEENKKM SQPGSPSPKT PTPSTPGDTQ
The product of this gene belongs to the SNF2/RAD54 helicase family. It represents the main component of the nucleosome remodeling and deacetylase complex and plays an important role in epigenetic transcriptional repression. Patients with dermatomyositis develop antibodies against this protein.Chemical Identifiers
| CHD4 | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 9530019N15Rik; AA617397; ATP-dependent helicase CHD4; BC005710; Chd4; CHD-4; chromodomain helicase DNA binding protein 4; chromodomain-helicase-DNA-binding protein 4; D6Ertd380e; Mi-2; Mi-2 autoantigen; Mi-2 autoantigen 218 kDa protein; Mi-2 beta; MI2B; Mi-2b; Mi-2beta; Mi2-BETA; mKIAA4075 | |
| Recombinant human CHD4 (immunogen range is 1291-1550). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CHD4 |
| K49015_15E2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Human | |
| Human | |
| Q14839 | |
| 1108 | |
| IgG1 | |
| Protein A | |
| Antibody |
Specifications
| CHD4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q14839 | |
| CHD4 | |
| Recombinant human CHD4 (immunogen range is 1291-1550). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| K49015_15E2 | |
| Unconjugated | |
| CHD4 | |
| 9530019N15Rik; AA617397; ATP-dependent helicase CHD4; BC005710; Chd4; CHD-4; chromodomain helicase DNA binding protein 4; chromodomain-helicase-DNA-binding protein 4; D6Ertd380e; Mi-2; Mi-2 autoantigen; Mi-2 autoantigen 218 kDa protein; Mi-2 beta; MI2B; Mi-2b; Mi-2beta; Mi2-BETA; mKIAA4075 | |
| Human | |
| Protein A | |
| RUO | |
| 1108 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |