missing translation for 'onlineSavingsMsg'
Learn More

CHCHD6 Monoclonal Antibody (K56003_20B7), Invitrogen™

Mouse Monoclonal Antibody

$1,557.73 - $2,745.56

Specifications

Antigen CHCHD6
Clone K56003_20B7
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA555691
View Documents
Invitrogen™
MA555691
50 μL N/A
Only null left
 
MA556071
View Documents
Invitrogen™
MA556071
200 μL N/A
Only null left
 
Description

Description

Sequence of this protein is as follows: RVSFGVDEEE RVRVLQGVRL SENVVNRMKE PSSPPPAPTS STFGLQDGNL RAPHKESTLP RSGSSGGQQP SGMKEGVKRY EQEHAAIQDK LFQVAKRERE AATKHSKASL PTGEGSISHE EQKSVRLARE LESREAELRR RDTFYKEQLE RIERKNAEMY KLSSEQFHEA ASKMESTIKP

CHCHD6 Monoclonal antibody specifically detects CHCHD6 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

CHCHD6
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
0710001P09Rik; 1700021B03Rik; CHCHD6; CHCM1; coiled coil helix cristae morphology 1; coiled coil helix cristae morphology protein 1; coiled-coil-helix cristae morphology 1; coiled-coil-helix cristae morphology protein 1; coiled-coil-helix-coiled-coil-helix domain containing 6; Coiled-coil-helix-coiled-coil-helix domain-containing protein 6; coiled-coil-helix-coiled-coil-helix domain-containing protein 6, mitochondrial; Mic25; MICOS complex subunit MIC25; PPP1R23; protein phosphatase 1, regulatory subunit 23
Recombinant human CHCHD6 (immunogen range is 11-190).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
CHCHD6
K56003_20B7
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse
Q91VN4, Q9BRQ6
66098, 84303
IgG1
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'