missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Charger for EVAHL (Hazardous Locations) PAPR Systems

Designed to be used with the Evolutionary Air Hazardous Location Powered Air Purifying Respirator (PAPR).

$635.01 - $3673.02

Specifications

Color Black
For Use With (Equipment) EVAHL PAPR Systems
View More Specs

Products 2
Catalog Number Mfr. No. No. of Ports Price Quantity  
Catalog Number Mfr. No. No. of Ports Price Quantity  
17-989-542
View Documents
Bullard™
EVAHLSMC
One
Each for $635.01
Only null left
 
17-989-544
View Documents
Bullard™
EVAHLGC
Six
Each for $3,673.02
Only null left
 
Description

Description

  • Choice of single or six port configurations to suit most needs

Specifications

Specifications

Black
EVAHL PAPR Systems
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'