missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bullard™ Charger for EVAHL (Hazardous Locations) PAPR Systems
Description
- Choice of single or six port configurations to suit most needs
Specifications
Specifications
| Color | Black |
| Product Type | Single-Unit Charging Station |
| For Use With (Equipment) | EVAHL PAPR Systems |
| Model | EVAHLSMC |
| No. of Ports | One |