missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ CHAPSO, Calbiochem™,

Catalog No. 2202021GM
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2202021GM 1 g Glass Bottle
2202025GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2202021GM Supplier MilliporeSigma™ Supplier No. 2202021GM

Chemical Identifiers

CAS 82473-24-3
Molecular Formula C32H58N2O8S
Molecular Weight (g/mol) 630.882
InChI Key GUQQBLRVXOUDTN-AKMBKZMMSA-N
Synonym 3-[(3-Cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate
PubChem CID 134129643
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8S,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]-2-hydroxypropane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CC(CS(=O)(=O)[O-])O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity >98% (TLC)