CAS RN 82473-24-3
IUPAC Name:
3-({3-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]propyl}dimethylazaniumyl)-2-hydroxypropane-1-sulfonate
Synonyms:
3-({3-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]propyl}dimethylammonio)-2-hydroxypropane-1-sulfonate
Molecular Weight (g/mol):
630.88
Molecular Formula:
C32H58N2O8S
InChi Key:
GUQQBLRVXOUDTN-XOHPMCGNSA-N
SMILES:
C[C@H](CCC(=O)NCCC[N+](C)(C)CC(O)CS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
1
–
1
of
1
results
1
MilliporeSigma™ CHAPSO, Calbiochem™,
CAS: 82473-24-3 Molecular Formula: C32H58N2O8S Molecular Weight (g/mol): 630.882 InChI Key: GUQQBLRVXOUDTN-AKMBKZMMSA-N Synonym: 3-[(3-Cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate PubChem CID: 134129643 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8S,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]-2-hydroxypropane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CC(CS(=O)(=O)[O-])O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C