Learn More
CHAPS, 98%, Thermo Scientific Chemicals
CHAPS, 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate, CAS # 75621-03-3, is a non-denaturing zwitterionic detergent that can be used for solubilizing membrane proteins.
Supplier: Thermo Scientific Chemicals 436550250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- CHAPS (3-((3-cholamidopropyl) dimethylammonio)-1-propanesulfonate) is a zwitterionic non-denaturing detergent
- It is suitable for applications which require protection of native protein conformation
Applications
- CHAPS can be used for the solubilization and purification of membrane proteins
- It is also used in conjunction with nonionic detergents like Triton X-100
- It can be used as a non-denaturing zwitterionic detergent for the study of membrane chemistry
Specifications
| CHAPS | |
| 267.2°C to 271.4°C | |
| 4.46 to 4.65 % N | |
| 98% | |
| C32H58N2O7S | |
| 25 g | |
| 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate | |
| UMCMPZBLKLEWAF-GBSSQDIOSA-N | |
| 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate | |
| 134129639 | |
| 98% |
| 75621-03-3 | |
| White | |
| Authentic | |
| Glass bottle | |
| MFCD00012116 | |
| 15,2043 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohols like methanol and ethanol | |
| CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C | |
| 614.9 | |
| 614.9 | |
| Crystalline Powder |
Chemical Identifiers
| 75621-03-3 | |
| 614.9 | |
| UMCMPZBLKLEWAF-GBSSQDIOSA-N | |
| 134129639 | |
| CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C |
| C32H58N2O7S | |
| MFCD00012116 | |
| 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate | |
| 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only