missing translation for 'onlineSavingsMsg'
Learn More

Chalcogran, 96%, mixture of isomers

Catalog No. AC461410010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461410010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC461410010 Supplier Thermo Scientific Chemicals Supplier No. 461410010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38401-84-2 | C9H16O2 | 156.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38401-84-2
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.22
InChI Key DCWKALWZHORJAO-UHFFFAOYSA-N
Synonym 2-ethyl-1,6-dioxaspiro 4.4 nonane, 2-ethyl-1,6-dioxaspiro 4.4-nonane, 2-ethyl-1,6-dioxaspiro 4.4 nonan, ethyl-1,6-dioxaspiro 4.4-nonane, 8-ethyl-4,9-dioxaspiro 4.4 nonane, 2-ethyl-1,6-dioxaspiro 4,4 nonane, 1,6-dioxaspiro 4.4 nonane, 2-ethyl, 2-ethyl-1,6-dioxaspiro 4.4 nonane #, 2-ethyl-1,6-dioxaspiro 4,4 nonane gc
PubChem CID 581383
IUPAC Name 2-ethyl-1,6-dioxaspiro[4.4]nonane
SMILES CCC1CCC2(O1)CCCO2

Specifications

Quantity 1 g
Boiling Point 67.0°C to 69.0°C
Packaging Glass Bottle
Assay Percent Range 95% min. (GC)
Formula Weight 156.22
Percent Purity 96%
Chemical Name or Material Chalcogran

RUO – Research Use Only