CAS RN 38401-84-2
IUPAC Name:
(2S,5R)-2-ethyl-1,6-dioxaspiro[4.4]nonane
Synonyms:
chalcogran
Molecular Weight (g/mol):
156.23
Molecular Formula:
C9H16O2
InChi Key:
DCWKALWZHORJAO-DTWKUNHWSA-N
SMILES:
CC[C@H]1CC[C@@]2(CCCO2)O1
1
–
1
of
1
results
1
Promotions Available
Chalcogran, 96%, mixture of isomers
Chalcogran, 96%, mixture of isomers
CAS: 38401-84-2 Molecular Formula: C9H16O2 Molecular Weight (g/mol): 156.22 InChI Key: DCWKALWZHORJAO-UHFFFAOYSA-N Synonym: 2-ethyl-1,6-dioxaspiro 4.4 nonane,2-ethyl-1,6-dioxaspiro 4.4-nonane,2-ethyl-1,6-dioxaspiro 4.4 nonan,ethyl-1,6-dioxaspiro 4.4-nonane,8-ethyl-4,9-dioxaspiro 4.4 nonane,2-ethyl-1,6-dioxaspiro 4,4 nonane,1,6-dioxaspiro 4.4 nonane, 2-ethyl,2-ethyl-1,6-dioxaspiro 4.4 nonane #,2-ethyl-1,6-dioxaspiro 4,4 nonane gc PubChem CID: 581383 IUPAC Name: 2-ethyl-1,6-dioxaspiro[4.4]nonane SMILES: CCC1CCC2(O1)CCCO2