missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGS 12066B dimaleate, 98+%, Thermo Scientific Chemicals
5-HT-1B full agonist
$229.49 - $229.49
Chemical Identifiers
| CAS | 109028-10-6 |
|---|---|
| Molecular Formula | C25H25F3N4O8 |
| Molecular Weight (g/mol) | 566.49 |
| MDL Number | MFCD00055048 |
| InChI Key | HTEVMLYDEWVIQE-LVEZLNDCSA-N |
| Synonym | cgs 12066b dimaleate, 1-methyl-4-7-trifluoromethyl pyrrolo 1,2-a quinoxalin-4-yl piperazine; bis butenedioic acid |
| PubChem CID | 71299720 |
| IUPAC Name | (E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline |
| SMILES | CN1CCN(CC1)C2=NC3=C(C=CC(=C3)C(F)(F)F)N4C2=CC=C4.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63801MA
|
Thermo Scientific Chemicals
J63801MA |
10 mg |
N/A
|
|
|||||
Description
5-HT-1B full agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 109028-10-6 | |
| 566.49 | |
| HTEVMLYDEWVIQE-LVEZLNDCSA-N | |
| 71299720 | |
| CN1CCN(CC1)C2=NC3=C(C=CC(=C3)C(F)(F)F)N4C2=CC=C4.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O |
| C25H25F3N4O8 | |
| MFCD00055048 | |
| cgs 12066b dimaleate, 1-methyl-4-7-trifluoromethyl pyrrolo 1,2-a quinoxalin-4-yl piperazine; bis butenedioic acid | |
| (E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline |
Specifications
| 109028-10-6 | |
| White | |
| MFCD00055048 | |
| cgs 12066b dimaleate, 1-methyl-4-7-trifluoromethyl pyrrolo 1,2-a quinoxalin-4-yl piperazine; bis butenedioic acid | |
| CN1CCN(CC1)C2=NC3=C(C=CC(=C3)C(F)(F)F)N4C2=CC=C4.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O | |
| 566.49 | |
| 566.49 | |
| Solid |
| 126°C to 127°C | |
| C25H25F3N4O8 | |
| 10 mg | |
| HTEVMLYDEWVIQE-LVEZLNDCSA-N | |
| (E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline | |
| 71299720 | |
| ≥98% | |
| CGS 12066B dimaleate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only