missing translation for 'onlineSavingsMsg'
Learn More

CGS 12066B dimaleate, 98+%, Thermo Scientific Chemicals

Catalog No. AAJ63801MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63801MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63801MA Supplier Thermo Scientific Chemicals Supplier No. J63801MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-HT-1B full agonist

5-HT-1B full agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109028-10-6
Molecular Formula C25H25F3N4O8
Molecular Weight (g/mol) 566.49
MDL Number MFCD00055048
InChI Key HTEVMLYDEWVIQE-LVEZLNDCSA-N
Synonym cgs 12066b dimaleate, 1-methyl-4-7-trifluoromethyl pyrrolo 1,2-a quinoxalin-4-yl piperazine; bis butenedioic acid
PubChem CID 71299720
IUPAC Name (E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline
SMILES CN1CCN(CC1)C2=NC3=C(C=CC(=C3)C(F)(F)F)N4C2=CC=C4.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 126°C to 127°C
Color White
Quantity 10 mg
Formula Weight 566.49
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material CGS 12066B dimaleate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only