Learn More
MilliporeSigma™ Supelco™ CGA-580 Gas Cylinder Pressure Regulators
Catalog No. 111008814
Change view
Click to view available options
Type:
1-Stage
2-Stage
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
111008814 1-Stage
111008548 2-Stage
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111008814 Supplier MilliporeSigma™ Supelco™ Supplier No. 29556U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Helium, Nitrogen, or Argon Gas Cylinders
Specifications
Type 1-Stage
For Use With (Equipment) Helium, Nitrogen, or Argon Gas Cylinders
Pressure 0 to 100 psi
Style High Purity