missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CF1 Mouse Embryonic Fibroblasts, MitC-treated

Mitomycin C (MitC)-treated Gibco CF1 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that are ideal for supporting healthy undifferentiated human and mouse embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) in culture.

$122.00

Specifications

Content And Storage Cryopreserved mouse embryonic fibroblasts

Store in liquid nitrogen.
Age Embryonic
Cell Type Feeder Cells (Fibroblast)
Form Cryopreserved
Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. No. of Cells Price Quantity & Availability  
Catalog Number Mfr. No. No. of Cells Price Quantity & Availability  
A34959
Encompass
View Documents Promotion Details
Gibco™
A34959
4 x 106
Each for $122.00
Only null left
 
A34958
Encompass
View Documents Promotion Details
Gibco™
A34958
2 x 106 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Manufactured by MTI-GlobalStem and equivalent to MTI-GlobalStem Cat. Nos. GSC-6001M and GSC-6201M, these widely-used MEF cells are isolated from outbred CF1 mice, mitotically arrested by MitC-treatment, and tested to ensure safety and performance.

• Help save precious time with pre-inactivated, ready-to-use MEFs
• Culture ESCs and iPSCs with confidence using meticulously tested feeder cells
• Rely on a widely-used product

Help save time with ready-to-use MEFs
Irradiated CF1 MEFs help save time and trouble dealing with an animal facility, performing tedious, time-consuming dissections, and expanding and testing cells day after day, week after week. Focus on your research with these pre-inactivated, ready-to-use feeders.

Culture with meticulously tested feeder cells
Each lot of CF1 MEFs is tested for bacteria, fungi, mycoplasma, and mouse pathogens to minimize the risk of contamination. Each lot is also tested for post-thaw viability (typically greater than 95%) and ability to support mouse and human ES cells to ensure robust and consistent performance. These MEFs have successfully been used with ESC/iPSC lines from other species, but should be tested for compatibility if used for such an application.

Rely on a widely-used product
These CF1 MEFs are used in many labs worldwide and have been cited in various publications. They have been successfully used to support undifferentiated pluripotent stem cells at densities ranging from 2 x 104–5.3 x 104 cells/cm2.

Specifications

Specifications

Cryopreserved mouse embryonic fibroblasts

Store in liquid nitrogen.
Feeder Cells (Fibroblast)
Mouse
Shipped on dry ice
Embryonic
Cryopreserved
Pooled
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.