missing translation for 'onlineSavingsMsg'
Learn More

Cetyltrimethylammonium bromide, MP Biomedicals™

Catalog No. ICN19502980
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19502980 100 g
ICN19502905 5 g
ICN19502990 500 g
ICN19502991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19502980 Supplier MP Biomedicals Inc Supplier No. 0219502980

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
Synonym Hexadecyltrimethylammonium bromide, CTAB
PubChem CID 5974
ChEBI CHEBI:3567
IUPAC Name hexadecyltrimethylazanium bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 0.9906
Formula Weight 364.5
Chemical Name or Material Cetyltrimethylammonium bromide
EINECSNumber 200-311-3
Recommended Storage Store at Room Temperature (15 to 30°C).