missing translation for 'onlineSavingsMsg'
Learn More

Cetyl Alcohol, NF, 90-102%, Spectrum™ Chemical

Catalog No. p-7157257
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601553 500 g Poly Bottle
18601554 2.5 kg Poly Pail
18601555 12 kg Fiber Drum
18601556 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601553 Supplier Spectrum Chemical Mfg Cor Supplier No. CE120500GM
Only null left

CE120, 36653-82-4, C16H34O

Spectrum™ Chemical Cetyl Alcohol, NF is used as an emulsion stabilizer, opacifier, emollient and surfactant. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 90 to 102%
Grade NF