missing translation for 'onlineSavingsMsg'
Learn More

Medisca Cetyl Alcohol NF

Catalog No. 14185633
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185633 1 kg
14185632 500 g
14185668 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14185633 Supplier Medisca Supplier No. 061809

Cetyl Alcohol NF comes as unctuous, white flakes, granules, cubes, or castings. It has a faint characteristic odor.

Cetyl Alcohol NF comes as unctuous, white flakes, granules, cubes, or castings. It has a faint characteristic odor.

Alternative name(s): 1-Cetanol, 1-Hexadecanol, Palmityl alcohol,

Specifications

Chemical Name or Material Cetyl Alcohol NF
Packaging Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck
CAS 36653-82-4
Type Drug Standard
Molecular Weight (g/mol) 242.44
Quantity 1 kg
Percent Purity 90 to 102 %
Grade NF