missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Cetyl Alcohol NF
Description
Cetyl Alcohol NF comes as unctuous, white flakes, granules, cubes, or castings. It has a faint characteristic odor.
Alternative name(s): 1-Cetanol, 1-Hexadecanol, Palmityl alcohol,
Specifications
Specifications
| Chemical Name or Material | Cetyl Alcohol NF |
| Packaging | Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 36653-82-4 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 242.44 |
| Quantity | 100 g |
| Percent Purity | 90 to 102 % |
| Grade | NF |