missing translation for 'onlineSavingsMsg'
Learn More

Cetyl alcohol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187458
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014045 1 g
111014046 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014045 Supplier Merck Emd Millipore Supplier No. 688241GF
Only null left

Cetyl alcohol is a long chain alcohol, which can find applications as an emulsifying agent, emollient, viscosity increasing agent, foam booster and opacifying agent in pharmaceuticals and cosmetics.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 36653-82-4
Flash Point 170°C
Physical Form Neat
Linear Formula CH3(CH2)15 OH
MDL Number MFCD00004760
Melting Point 49°C to 51°C
Boiling Point 190°C (15 mmHg)
Synonym 1-Hexadecanol; Palmityl alcohol
Type Analytical Standard
Quantity 1 g
Formula Weight 242.44
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less