missing translation for 'onlineSavingsMsg'
Learn More

Cetraxate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7125964
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3454250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3454250MG Supplier TCI America Supplier No. C3454250MG

Chemical Identifiers

CAS 27724-96-5
Molecular Formula C17H24ClNO4
Molecular Weight (g/mol) 341.83
MDL Number MFCD01722166
InChI Key USROQQUKEBHOFF-UHFFFAOYSA-N
Synonym 3-[4-[[trans-4-(Aminomethyl)cyclohexyl]carbonyloxy]phenyl]propanoic Acid Hydrochloride
PubChem CID 34001
IUPAC Name hydrogen 3-{4-[4-(aminomethyl)cyclohexanecarbonyloxy]phenyl}propanoic acid chloride
SMILES [H+].[Cl-].NCC1CCC(CC1)C(=O)OC1=CC=C(CCC(O)=O)C=C1

Specifications

Melting Point 240°C
Color White
Quantity 250 mg
Formula Weight 341.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cetraxate Hydrochloride