missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine hydrochloride Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013687
Change view
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013687 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013687 Supplier Merck Emd Millipore Supplier No. PHR171040MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cetirizine hydrochloride Related Compound A is an impurity of cetirizine hydrochloride. Cetirizine hydrochloride is a second-generation antihistamine compound. It is a major metabolite of hydroxyzine that exhibits high specific affinity for histamine H1 receptor. It is used for the management of allergies, hay fever, angioedema, and urticaria.Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 246870-46-2 (free base)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H29 ClN2O3 · 2 HCl
Synonym Cetirizine ethyl ester dihydrochloride; (RS)-2-[2-[4-[(4-chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic acid ethyl ester dihydrochloride (Cetirizine hydrochloride Related Compound A); (RS)-2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic acid ethyl ester dihydrochloride
Type Certified Reference Material
Quantity 40 mg
Formula Weight 489.86
Grade Certified Reference Material