missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetirizine Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America C231625G
Specifications
| Cetirizine Dihydrochloride | |
| 225°C | |
| C21H27Cl3N2O3 | |
| 25 g | |
| PGLIUCLTXOYQMV-UHFFFAOYNA-N | |
| dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride | |
| 55182 | |
| ≥98.0% (HPLC,T) |
| 83881-52-1 | |
| White | |
| MFCD00941428 | |
| cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler | |
| [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 | |
| 461.81 | |
| 461.81 | |
| Crystalline Powder |
Chemical Identifiers
| 83881-52-1 | |
| 461.81 | |
| PGLIUCLTXOYQMV-UHFFFAOYNA-N | |
| 55182 | |
| [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 |
| C21H27Cl3N2O3 | |
| MFCD00941428 | |
| cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler | |
| dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride |
Safety and Handling
RTECSNumber : AG0980000
TSCA : No