Learn More
Cetirizine dihydrochloride, 99+%
CAS: 83881-52-1 | C21H27Cl3N2O3 | 461.81 g/mol
Supplier: Thermo Scientific Chemicals J6354914
Description
Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.
Solubility
Soluble in water, DMSO, ethanol, and methanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
| Cetirizine Dihydrochloride | |
| White | |
| MFCD00941428 | |
| 14,2022 | |
| Soluble in water,DMSO,ethanol,and methanol. | |
| [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 | |
| 461.81 | |
| 461.82 | |
| Solid |
| 83881-52-1 | |
| C21H27Cl3N2O3 | |
| 25 g | |
| cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler | |
| PGLIUCLTXOYQMV-UHFFFAOYNA-N | |
| dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride | |
| 55182 | |
| ≥99% |
Chemical Identifiers
| 83881-52-1 | |
| 461.81 | |
| PGLIUCLTXOYQMV-UHFFFAOYNA-N | |
| 55182 | |
| [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 |
| C21H27Cl3N2O3 | |
| MFCD00941428 | |
| cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler | |
| dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
RTECSNumber : AG0980000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only