Learn More
Cesium sulfate, 99+%, pure
CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol
Supplier: Thermo Scientific Chemicals 218700250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1900.0°C | |
| 1005.0°C | |
| 10294-54-9 | |
| Fine Crystalline Powder | |
| 99% min | |
| Cs2SO4 | |
| 15,2015 | |
| Solubility in water: 1820g/L (25°C). Other solubilities: practically insoluble in alcohol, aceton, pyridine | |
| [Cs+].[Cs+].[O-]S([O-])(=O)=O | |
| 361.87 | |
| 361.86 | |
| Pure | |
| 4.24 |
| Cesium sulfate | |
| 99+% | |
| White | |
| 25 g | |
| Cs2O4S | |
| MFCD00010959 | |
| cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate | |
| FLJPGEWQYJVDPF-UHFFFAOYSA-L | |
| dicaesium(1+) sulfate | |
| 25137 | |
| ≥99% | |
| Glass bottle |
Chemical Identifiers
| 10294-54-9 | |
| 361.87 | |
| FLJPGEWQYJVDPF-UHFFFAOYSA-L | |
| 25137 |
| Cs2O4S | |
| MFCD00010959 | |
| cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate | |
| [Cs+].[Cs+].[O-]S([O-])(=O)=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
Obtain special instructions before us
GHS Signal Word: Warning
EINECSNumber : 233-662-6
RTECSNumber : FL0800000
TSCA : TSCA
RUO – Research Use Only