missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium sulfate, 99+%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1900.0°C |
| Melting Point | 1005.0°C |
| Color | White |
| Physical Form | Fine Crystalline Powder |
| Quantity | 250 g |
| Assay Percent Range | 99% min |
| Linear Formula | Cs2SO4 |
| Merck Index | 15,2015 |
| Solubility Information | Solubility in water: 1820g/L (25°C). Other solubilities: practically insoluble in alcohol, aceton, pyridine |
| IUPAC Name | dicaesium(1+) sulfate |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wash face,hands and any exposed skin thoroughly after handling. Call a POISON CENTER or doctor/physician if you feel unwell. Obtain special instructions before us |
| EINECSNumber | 233-662-6 |
| RTECSNumber | FL0800000 |
| TSCA | TSCA |
RUO – Research Use Only