missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, 99+%, pure

Catalog No. AC218700100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218700100 10 g Glass bottle
AC218700250 25 g Glass bottle
AC218701000 100 g Glass bottle
AC218702500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC218700100 Supplier Thermo Scientific Chemicals Supplier No. 218700100
Only null left

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
PubChem CID 25137
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1900.0°C
Melting Point 1005.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 10 g
Assay Percent Range 99% min
Linear Formula Cs2SO4
Merck Index 15,2015
Solubility Information Solubility in water: 1820g/L (25°C). Other solubilities: practically insoluble in alcohol, aceton, pyridine
IUPAC Name dicesium;sulfate
Formula Weight 361.86
Percent Purity ≥99%
Grade Pure
Packaging Glass bottle
Density 4.24
Chemical Name or Material Cesium sulfate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
Obtain special instructions before us
EINECSNumber 233-662-6
RTECSNumber FL0800000
TSCA TSCA

RUO – Research Use Only