missing translation for 'onlineSavingsMsg'
Learn More

Cesium Nitrate, 99%+, MilliporeSigma™

Catalog No. p-4939035
Change view
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1028560025 25 g Glass bottle
M1028561000 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1028560025 Supplier Technidata Supplier No. 1.02856.0025
Only null left

Chemical Identifiers

CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
Synonym cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Physical Form Solid
Quantity 25 g
Assay Percent Range 99%+
UN Number UN 1451
Formula Weight 194.91
Odor Odorless
Packaging Glass bottle
Appearance Off-white solid
Chemical Name or Material Cesium nitrate
Hazard Category H272: May intensify fire; oxidiser.
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 232-146-8