missing translation for 'onlineSavingsMsg'
Learn More

Cesium nitrate, 99.9%, (trace metal basis), for analysis

Catalog No. p-3734721
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC192070500 50 g Glass bottle
AC192072500 250 g Glass bottle
AC192070010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC192070500 Supplier Thermo Scientific Chemicals Supplier No. 192070500
Only null left

CAS: 7789-18-6 | CsNO3 | 194.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.91
MDL Number MFCD00010963
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
Synonym cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Melting Point 414.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 50 g
Assay Percent Range 99.9%
Linear Formula CsNO3
Merck Index 15,2014
Solubility Information Solubility in water: 9.16 g/100mL (0°C). Other solubilities: soluble in acetone, slightly soluble in alcohol
Formula Weight 194.91
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 3.68
Density 3.6800g/mL
Chemical Name or Material Cesium nitrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 232-146-8

RUO – Research Use Only