missing translation for 'onlineSavingsMsg'
Learn More

Cesium methanesulfonate, 98%

Catalog No. p-4258679
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431580050 5 g Glass bottle
AC431580250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431580050 Supplier Thermo Scientific Chemicals Supplier No. 431580050
Only null left

CAS: 2550-61-0 | CH3CsO3S | 228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2550-61-0
Molecular Formula CH3CsO3S
Molecular Weight (g/mol) 228
InChI Key DJKJXRLREATOMF-UHFFFAOYSA-M
Synonym cesium methanesulfonate, csmes, csmsf, methanesulfonic acid cesium salt, caesium 1+ ion mesylate, cesium methanesulfonate used in patch clamp techniques, cesium methanesulfonate, used in patch clamp techniques
PubChem CID 5148066
IUPAC Name cesium;methanesulfonate
SMILES CS(=O)(=O)[O-].[Cs+]

Specifications

Melting Point 262.0°C to 264.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 98%
Solubility Information (5% in water) Clear colorless
Formula Weight 228
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Cesium methanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only