missing translation for 'onlineSavingsMsg'
Learn More

Cesium iodide for high-reSolution mass spectroscopy, 0.9999, MilliporeSigma™ Supelco™

Catalog No. 111020168
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020168 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020168 Supplier Merck Emd Millipore Supplier No. 210041G
Only null left

Cesium iodide is a simple ionic salt. It has the capacity to undergo second order transformation from cubic B2 structure to body-centered tetragonal structure at low pressures. It is mostly used as input screens of X-ray image intensifiers.

Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

CAS 7789-17-5
Density 4.51 g/mL (at 25°C)
Physical Form Neat
Linear Formula CsI
MDL Number MFCD00010962
Melting Point 626°C
Type Analytical Standard
Quantity 1 g
Formula Weight 259.81
Percent Purity 99.99%
Grade Analytical Standard
Show More Show Less