missing translation for 'onlineSavingsMsg'
Learn More

Cesium formate, 98%

Catalog No. AAB2132714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2132714 25 g
AAB2132722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2132714 Supplier Thermo Scientific Chemicals Supplier No. B2132714
Only null left

CAS: 3495-36-1 | CH2CsO2 | 178.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3495-36-1
Molecular Formula CH2CsO2
Molecular Weight (g/mol) 178.93
MDL Number MFCD00149883
InChI Key ATZQZZAXOPPAAQ-UHFFFAOYSA-N
Synonym cesium formate, formic acid, cesium salt, formic acid, cesium salt 1:1, formic acid cesium salt, 64-18-6 parent, cesuim formate, acmc-1ajhs, cho2.cs, caesium 1+ formate
PubChem CID 23663999
IUPAC Name caesium(1+) formic acid
SMILES [Cs+].OC=O

Specifications

Quantity 25 g
Beilstein 3912426
Formula Weight 177.92
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Cesium formate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 222-492-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only