accessibility menu, dialog, popup

UserName

CsHOO

CAS RN 3495-36-1

IUPAC Name: caesium(1+) formic acid
Synonyms: caesium(1+) formic acid
Molecular Weight (g/mol): 178.93
Molecular Formula: CH2CsO2
InChi Key: ATZQZZAXOPPAAQ-UHFFFAOYSA-N
SMILES: [Cs+].OC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results