missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, TraceSELECT™, ≥99.99% (Metals Basis), Honeywell Fluka™

TraceSELECT™, for trace analysis,≥99.9995% (metals basis)

$1545.05 - $1545.05

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60047737
View Documents
Honeywell Chemicals
90033100G
100 g Poly Bottle
Each for $1,545.05
Only null left
 
Description

Description

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Specifications

Chemical Identifiers

7647-17-8
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
cesium;chloride
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]

Specifications

642°C
Colorless
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36g/mol
TraceSELECT
Poly Bottle
3.98g/cm3 at 20°C
7647-17-8
100 g
CsCl
NONH for all modes of transport
In water 1000g/L at 20°C
[Cl-].[Cs+]
168.36
CHEBI:63039
≥99.9995% (metals basis)
Odorless
Powder
Cesium chloride
Videos
Safety and Handling

Safety and Handling

P280

RTECSNumber : 231-600-2

SDS
missing translation for 'documents'

missing translation for 'documents'