missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, TraceSELECT™, ≥99.99% (Metals Basis), Honeywell Fluka™

Catalog No. 60047737
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047737 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60047737 Supplier Honeywell Chemicals Supplier No. 90033100G
Only null left

TraceSELECT™, for trace analysis,≥99.9995% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]

Specifications

Melting Point 642°C
Color Colorless
Quantity 100 g
Linear Formula CsCl
UN Number NONH for all modes of transport
Solubility Information In water 1000g/L at 20°C
Formula Weight 168.36g/mol
Percent Purity ≥99.9995% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly Bottle
Appearance Powder
Density 3.98g/cm3 at 20°C
Chemical Name or Material Cesium chloride
Show More Show Less
Hazard Statement P280
RTECSNumber 231-600-2