missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, 99.0%, (Reagent Grade), MP Biomedicals

$114.70 - $1723.99

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10132125
View Documents
MP Biomedicals Inc
0210132125
25 g
Each for $114.70
Only null left
 
ICN10132180
View Documents
MP Biomedicals Inc
0210132180
100 g
Each for $324.71
Only null left
 
ICN10132183
View Documents
MP Biomedicals Inc
0210132183
250 g
Each for $585.86
Only null left
 
ICN10132190
View Documents
MP Biomedicals Inc
0210132190
500 g
Each for $1,072.94
Only null left
 
ICN10132191
View Documents
MP Biomedicals Inc
0210132191
1 kg
Each for $1,723.99
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7647-17-8
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
[Cl-].[Cs+]
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039

Specifications

7647-17-8
99.0%
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
caesium(1+) chloride
24293
168.4
Cesium Chloride
25 g
ClCs
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
[Cl-].[Cs+]
168.36
CHEBI:63039
Reagent
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'