missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride, 99.0%, (Reagent Grade), MP Biomedicals
Supplier: MP Biomedicals Inc 0210132183
Specifications
| Cesium Chloride | |
| 250 g | |
| ClCs | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| [Cl-].[Cs+] | |
| 168.36 | |
| CHEBI:63039 | |
| Reagent |
| 7647-17-8 | |
| 99.0% | |
| MFCD00010955 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| cesium;chloride | |
| 24293 | |
| 168.4 |
Chemical Identifiers
| 7647-17-8 | |
| 168.36 | |
| AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
| 24293 | |
| [Cl-].[Cs+] |
| ClCs | |
| MFCD00010955 | |
| cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
| CHEBI:63039 |
Safety and Handling
Recommended Storage : Room Temperature