missing translation for 'onlineSavingsMsg'
Learn More

Cesium Carbonate 98.0+%, TCI America™

Catalog No. p-7125310
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C216025G 25 g
C2160100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C216025G Supplier TCI America Supplier No. C216025G
Only null left

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
Synonym cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
PubChem CID 10796
IUPAC Name dicaesium(1+) carbonate
SMILES [Cs+].[Cs+].[O-]C([O-])=O

Specifications

Melting Point 610°C
Color White
pH 10 to 12 (50 g/L at 20°C)
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 325.82
Percent Purity ≥98.0% (T)
Chemical Name or Material Cesium Carbonate
RTECSNumber FK9400000
TSCA Yes