missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Carbonate 98.0+%, TCI America™
Description
- Suitable for laboratory applications
Specifications
Specifications
| Melting Point | 610°C |
| Color | White |
| pH | 10 to 12 (50 g/L at 20°C) |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Assay Percent Range | >98.0% |
| Merck Index | 2010 |
| Solubility Information | 2.77 g/L in Water |
| Formula Weight | 325.82 |
| Percent Purity | ≥98.0% (T) |
| Show More |
Safety and Handling
| EINECSNumber | (1)-1061 |
| RTECSNumber | FK9400000 |
| TSCA | Yes |