missing translation for 'onlineSavingsMsg'
Learn More

Cesium carbonate, 99% (metals basis)

Catalog No. p-4407363
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1288706 5 g
AA1288718 50 g
AA1288730 250 g
AA12887A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1288706 Supplier Thermo Scientific Chemicals Supplier No. 01288706
Only null left

Catalyst in ethylene oxide polymerization, in coating spatter-free welding of steel in CO2, and in oxide cathodes | CAS: 534-17-8 | CCs2O3 | 325.82 g/mol

Cesium carbonate is the precursor for other cesium compounds. It acts as a base in sensitive organic reactions. Suzuki, Heck and Sonogashira synthesis are some of the important reactions involving cesium carbonate. It finds use in solar cells as it increases the power conversion efficiency of cells through the transfer of electrons. It is also used in the production of special optical glasses, petroleum catalytic additives, special ceramics, and in sulfuric acid industry. It is useful in the N-alkylation (of sulfonamides, beta-lactams, indoles, heterocycles, and several sensitive nitrogen compounds), carbamination of amines, carbonylation of alcohols, and aerobic oxidation of alcohols into carbonyl compounds without over oxidations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
Synonym cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O

Specifications

Melting Point 610°C (decomposition)
Physical Form Powder/Granules
Quantity 5 g
Assay Percent Range 99% (metals basis)
Beilstein 4546405
Solubility Information Freely soluble in water. Soluble in ethanol,dimethylformamide and dimethyl sulfoxide.
IUPAC Name dicaesium(1+) carbonate
Formula Weight 325.82
Sensitivity Hygroscopic
Density 4.072 g/mL
Chemical Name or Material Cesium carbonate
Show More Show Less
Hazard Category H318-H361f-H373
Precautionary Statement P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c
EINECSNumber 208-591-9
RTECSNumber FK9400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only