missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CES7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113939 Supplier Invitrogen™ Supplier No. PA5113939
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: SGNWVHPGQI LIWAIWVLAA PTKGPSAEGP QRNTRLGWIQ GKQVTVLGSP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs.

Specifications

Antigen CES7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CES5A
Gene Accession No. Q6NT32
Gene Alias 1700081L16Rik; 1700122C07Rik; BB081581; carboxylesterase 5; carboxylesterase 5A; carboxylesterase 7; carboxylesterase-like urinary excreted protein homolog; CAUXIN; CES4C1; CES5; Ces5a; CES7; epididymis luminal protein 126; Gm503; HEL126
Gene Symbols CES5A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human CES7 (aa 2-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221223
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less