missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Certipur™ UV/VIS standards

Catalog No. p-4938800
Change view
Click to view available options
Type:
UV-VIS Standard 1: Potassium dichromate solution
UV-VIS Standard 1A: Potassium dichromate solution
UV-VIS Standard 2: Sodium nitrite solution
UV-VIS Standard 4: Potassium chloride solution
UV-VIS Standard 5: Toluene solution in n-hexane
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
M1081640001 UV-VIS Standard 4: Potassium chloride solution
M1081610001 UV-VIS Standard 2: Sodium nitrite solution
M1046600001 UV-VIS Standard 1A: Potassium dichromate solution
M1081650001 UV-VIS Standard 5: Toluene solution in n-hexane
M1081600001 UV-VIS Standard 1: Potassium dichromate solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M1081640001 Supplier MilliporeSigma™ Supplier No. 1.08164.0001
Only null left