accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
Environmental Testing Standards
Environmental Chemical Standards
MilliporeSigma™ Certipur™ UV/VIS standards
Print
MilliporeSigma™ Certipur™ UV/VIS standards
Catalog No.
M1081640001
Change view
Chip view icon
List view icon
Click to view available options
Type:
UV-VIS Standard 1: Potassium dichromate solution
UV-VIS Standard 1A: Potassium dichromate solution
UV-VIS Standard 2: Sodium nitrite solution
UV-VIS Standard 4: Potassium chloride solution
UV-VIS Standard 5: Toluene solution in n-hexane
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Type
M1081640001
UV-VIS Standard 4: Potassium chloride solution
M1081610001
UV-VIS Standard 2: Sodium nitrite solution
M1046600001
UV-VIS Standard 1A: Potassium dichromate solution
M1081650001
UV-VIS Standard 5: Toluene solution in n-hexane
M1081600001
UV-VIS Standard 1: Potassium dichromate solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No.
M1081640001
Supplier
MilliporeSigma™
Supplier No.
1.08164.0001
Sign In
or
Register
to check your price and availability.
$1,422.54
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Description
For Qualification of a UV/VIS spectrometer
Specifications
Specifications
Type
UV-VIS Standard 4: Potassium chloride solution
Product Suggestions
Documents